Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver, brazing alloy, flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long

Manufacturer:  Alfa Aesar™ 41031KG

Quantity 1 piece

 View more versions of this product

Catalog No. AA41031KG


  Check Availability
Add to cart

Specifications

Description

  • Low temperature alloy for use on ferrous and nonferrous material; good color match for stainless material
Description & Specifications

Chemical Identifiers

MFCD00801120
Silver, brazing Alloy, Flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long, Tensile Strength: 79,000 psi

Specifications

Ag:Cu:Zn:Sn; 56:22:17:5 wt% Nontoxic, cadmium free
1 piece
MFCD00801120
Silver, brazing Alloy, Flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long, Tensile Strength: 79,000 psi

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit