Alfa Aesar™ Silver, brazing flux, white paste

Manufacturer: Alfa Aesar™ 41355TB

Quantity 113g

 View more versions of this product

Catalog No. AA41355TB

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD01868499
    Chemical Name or Material Silver, brazing flux, Active range: 540 to 980°

    Specifications

    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Paste
    MDL Number MFCD01868499
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H360-H315-H319-H335
    May damage fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color White
    Quantity 113g
    Chemical Name or Material Silver, brazing flux, Active range: 540 to 980°

    GHS H Statement
    H360-H315-H319-H335
    May damage fertility or the unborn child.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit