Learn More
Silver carbonate on Celite, ≈0.7 mmole Ag2CO3/g reagent, Thermo Scientific Chemicals
Description
Silver carbonate is used in production of silver powder. It is used in microelectronics. Silver carbonate is used as a reagent in organic synthesis. It is employed in Fetizon oxidation, silver carbonate on celite acts as an oxidizing agent to form lactones from diols. It is also used in used in Koenigs-Knorr glycosylation. It is also employed to convert alkyl bromides into alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Powder |
Quantity | 15 mmol |
Solubility Information | Soluble in water (0.03mg/ml at 20°C),dilute HNO3,ammonia and ammonium hydroxide. Insoluble in alcohol. |
Odor | Odorless |
Sensitivity | Light sensitive |
Chemical Name or Material | Silver carbonate on Celite, ∼0.7 mmole Ag2CO3/g reagent |
Safety and Handling
Hazard Category | H318-H335-H373 |
Hazard Statement | GHS H Statement H351-H373-H319 Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Causes serious eye irritation. |
Precautionary Statement | P260-P271-P280-P304+P340-P305+P351+P338-P310-P312-P314-P501c |
DOTInformation | Hazard Class: 9 |
EINECSNumber | 208-590-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.