Alfa Aesar™ Silver chloride, 4.9% in H2O, nanoparticle dispersion, <200nm APS Number Weighted

Manufacturer: Alfa Aesar™ 4728022

Quantity 100g

 View more versions of this product

Catalog No. AA4728022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-90-6
    Formula Weight 143.32
    MDL Number MFCD00003399
    UN Number UN3082
    Merck Index 14,8509
    Synonym Silver chloride dispersion in water
    Chemical Name or Material Silver chloride, Nanoparticle dispersion

    Specifications

    Formula Weight 143.32
    Concentration 4.9% in H2O
    UN Number UN3082
    Odor Odorless
    Particle Size <200nm APS Number Weighted
    Density 1.038g/mL
    Physical Form Liquid
    Synonym Silver chloride dispersion in water
    Sensitivity Light Sensitive
    Storage Note 1 2° to 8°C
    DOT Information Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
    Quantity 100g
    TSCA TSCA
    MDL Number MFCD00003399
    CAS 7783-90-6
    Merck Index 14,8509
    Color White opaque
    RTECS Number VW3563000
    Chemical Name or Material Silver chloride, Nanoparticle dispersion

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit