Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver chloride, 99.9% (metals basis), Alfa Aesar™

Click to view available options
Quantity:
25g
100g
500g
7783-90-6
AgCl
MFCD00003399
7783-90-6
AgCl
MFCD00003399
Assay Percent Range 99.9% (metals basis)
Appearance White Powder/crystalline/beads
Solubility Information Practically insoluble in water. Soluble in ammonia,solutions of alkali cyanides,thiosulfates and ammonium carbonates. Insoluble in alcohol and dilute acids.
Formula Weight 143.32
Physical Form Crystalline
Density 5.56
Odor Odorless
Refractive Index 2.071
Sensitivity Light sensitive
Boiling Point 1550°C
Color Yellow
Melting Point 455°C
Quantity 500g
Chemical Name or Material Silver chloride
Show More Show Less

P234-P390

H290

DOTInformation : DOT Class: 8; Packaging Group: III

EINECSNumber : 232-033-3

RTECSNumber : VW3563000

TSCA : Yes

Recommended Storage : Ambient temperatures