Alfa Aesar™ Silver cyclohexanebutyrate

Manufacturer: Alfa Aesar™ 4370803

Quantity 1g

 View more versions of this product

Catalog No. AA4370803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62638-04-4
    Molecular Formula C10H17AgO2
    Formula Weight 277.11
    MDL Number MFCD00036396
    Synonym Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
    Chemical Name or Material Silver cyclohexanebutyrate

    Specifications

    Formula Weight 277.11
    Molecular Formula C10H17AgO2
    CAS 62638-04-4
    Synonym Cyclohexanebutyric acid silver salt; 4-Cyclohexylbutyric acid silver salt
    TSCA TSCA
    Sensitivity Light Sensitive
    Melting Point ≈255°C (dec.)
    Physical Form Powder
    MDL Number MFCD00036396
    Quantity 1g
    Chemical Name or Material Silver cyclohexanebutyrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit