Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver diethyldithiocarbamate, ACS, Thermo Scientific Chemicals

Catalog Number AA1189809
Click to view available options
Quantity:
10 g
50 g
1470-61-7
C5H10AgNS2
256.13
MFCD00004929
NSVHDIYWJVLAGH-UHFFFAOYSA-M
silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
3034078
silver;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].[Ag+]
1470-61-7
C5H10AgNS2
256.13
MFCD00004929
NSVHDIYWJVLAGH-UHFFFAOYSA-M
silver diethyldithiocarbamate, unii-gvn001b86o, agddtc, silver n,n-diethylcarbamodithioate, diethyldithiocarbamic acid, silver salt, diethyl dithio carbamic acid silver salt, silver i diethyldithiocarbamate, silver, diethylcarbamodithioato-ks,ks', ditiocarb silver, silver n,n-diethyldithiocarbamate
3034078
silver;N,N-diethylcarbamodithioate
CCN(CC)C(=S)[S-].[Ag+]

Silver diethyldithiocarbamate cluster plays a useful role for the deposition of acanthite (Ag2S) thin films which is employed in photoelectrochemical applications. Silver diethyldithiocarbamate membrane is used to develop ion selective electrodes for nitrate ion. It is useful in the colorimetric determination of arsenic. It acts as a copper chelator, and is utilized to quantitate copper in biological materials by ESR. It also works as an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 172°C to 175°C
Color White
Physical Form Powder
Quantity 10 g
Solubility Information It is soluble in water and pyridine.
Formula Weight 256.14
Grade ACS
Odor Odorless
Sensitivity Light sensitive
Appearance Yellow to pale brown powder
Chemical Name or Material Silver diethyldithiocarbamate, For analysis ACS
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-003-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit