Learn More
MP Biomedicals™ Silver Enhancing Kit, 2x250mL
Used for amplifying the signal from gold particles attached to proteins or antigens immobilized on membranes
Manufacturer:  MP Biomedicals™ 08678211
Catalog No. ICN678211
Add to cart
Specifications
Specifications
Immunoblotting, Immunoassays
2° to 8°C
Silver Enhancing Kit
2 x 250 mL