Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Foil, 0.25mm (0.01 in.) thick, hard, Premion™, 99.998% (Metals basis), Alfa Aesar™

Click to view available options
Quantity:
50 x 50mm
100 x 100mm
100 x 500mm
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]

Form: Approx. 6.6g/50 x 50mm; Temper: as rolled

Quantity 50 x 50mm
Appearance Silver-colored foil
Density 10.5g/cm3
Assay Percent Range 99.998% (Metals basis)
Odor Characteristic
Physical Form Foil, 0.25mm (0.01 in.) thick
Color Silver
Chemical Name or Material Silver, Hard

EINECSNumber : 231-131-3

TSCA : Yes

Recommended Storage : Ambient temperatures