Silver Foil, 1.0mm (0.04 in.) thick, Premion™, 99.9985% (Metals basis), Alfa Aesar™

Form: Approx. 6.6g/25 x 25mm

Manufacturer: Alfa Aesar 12127GH

Quantity 100 x 100mm

 View more versions of this product

Catalog No. AA12127GH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-22-4
    Physical Form Foil, 1.0mm (0.04 in.) thick
    Assay Percent Range 99.9985% (Metals basis)
    MDL Number MFCD00003397
    Chemical Name or Material Silver

    Specifications

    Quantity 100 x 100mm
    Color Silver
    Physical Form Foil, 1.0mm (0.04 in.) thick
    Assay Percent Range 99.9985% (Metals basis)
    MDL Number MFCD00003397
    CAS 7440-22-4
    Odor Characteristic
    Appearance Silver-colored foil
    Chemical Name or Material Silver

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit