Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver Gauze, 80 mesh woven from 0.115mm (0.0045 in.) dia. wire, 99.9% (Metals basis), Alfa Aesar™
Click to view available options
Quantity:
25 x 25mm
75 x 75mm
150 x 150mm
300 x 300mm
Specifications
Quantity | 75 x 75mm |
Appearance | Silver-colored guaze |
Assay Percent Range | 99.9% (Metals basis) |
Mesh Size | 80 Mesh |
Solubility Information | Insoluble in water. |
Odor | Characteristic |
Physical Form | Wire Cloth |
Color | Silver |
Chemical Name or Material | Silver |
Safety and Handling
EINECSNumber : 231-131-3
TSCA : Yes
Recommended Storage : Ambient temperatures