Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver hexafluorophosphate, 98%, -6 mesh, ACROS Organics™

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Plastic bottle
26042-63-7
AgF6P
252.83
MFCD00003415
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
168464
silver;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Ag+]
26042-63-7
AgF6P
252.83
MFCD00003415
SCQBROMTFBBDHF-UHFFFAOYSA-N
silver hexafluorophosphate, silver hexafluorophosphate v, ag.f6p, silver i hexafluorophosphate, agpf6, acmc-1ceef, phosphate 1-, hexafluoro-, silver 1+, phosphate 1-, hexafluoro-, silver 1+ 1:1, silver i hexafluorophosphate v
168464
silver;hexafluorophosphate
F[P-](F)(F)(F)(F)F.[Ag+]
Linear Formula AgPF6
Solubility Information Solubility in water: soluble
Formula Weight 252.83
Physical Form Powder
Percent Purity 98%
Packaging Plastic bottle
Color Beige-Gray to White
Quantity 1g
Chemical Name or Material Silver hexafluorophosphate

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 247-428-6