Learn More
Silver hexafluorophosphate, 98%, Thermo Scientific Chemicals
Description
It is commonly used as an oxidant forming silver metal. It is often used as a reagent in inorganic and organometallic chemistry. It is used in the preparation of engineering novel materials as well as in preparation of supra molecular Ag(I) complexes. It is used in the development of new catalysts, replacing halide ligands with the weakly coordinating hexafluorophosphate anion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 102°C (decomposition) |
Color | Brown to Yellow |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in organic solvents such as benzene,toluene,and acetonitrile. |
Formula Weight | 252.83 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | White powder |
Show More |
Safety and Handling
Hazard Category | H314 |
Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8; Packaging Group: II |
EINECSNumber | 247-428-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.