Alfa Aesar™ Silver hydrogen fluoride, 99% min

Manufacturer: Alfa Aesar™ 4012918

Quantity 50g

 View more versions of this product

Catalog No. AA4012918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12249-52-4
    Linear Formula AgHF2
    Formula Weight 146.87
    MDL Number MFCD00042145
    UN Number UN1740
    Synonym Silver bifluoride
    Chemical Name or Material Silver hydrogen fluoride

    Specifications

    Formula Weight 146.87
    Health Hazard 3 GHS P Statement
    P280-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Linear Formula AgHF2
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Physical Form Crystalline
    Synonym Silver bifluoride
    Quantity 50g
    Health Hazard 1 Danger
    Sensitivity Light Sensitive & Hygroscopic
    MDL Number MFCD00042145
    CAS 12249-52-4
    UN Number UN1740
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROGENDIFLUORIDE, SOLID, N.O.S.
    Chemical Name or Material Silver hydrogen fluoride
    Assay ≤99%

    GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P280-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit