Alfa Aesar™ Silver(I) chromate, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4549514

Quantity 25g

 View more versions of this product

Catalog No. AA4549514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7784-01-2
    Molecular Formula O4Ag2Cr
    Linear Formula Ag2CrO4
    Formula Weight 331.73
    MDL Number MFCD00003402
    UN Number UN1479
    Merck Index 14,8510
    Chemical Name or Material Silver (I) chromate

    Specifications

    Formula Weight 331.73
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Odor Odorless
    Molecular Formula O4Ag2Cr
    CAS 7784-01-2
    UN Number UN1479
    Health Hazard 1 Danger
    Physical Form Powder
    Linear Formula Ag2CrO4
    MDL Number MFCD00003402
    Merck Index 14,8510
    Color Red-brown
    Health Hazard 2 GHS H Statement
    H350-H272-H317
    May cause cancer.
    May intensify fire
    oxidizer.
    May cause an allergic skin reaction.
    Density 5.63g/mL
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Sensitivity Light Sensitive
    Quantity 25g
    Chemical Name or Material Silver (I) chromate
    Assay 99.9%, (metals basis)

    GHS H Statement
    H350-H272-H317
    May cause cancer.
    May intensify fire
    oxidizer.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P220-P363-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    Wash contaminated clothing before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit