Learn More
Silver(I) fluoride, 98%, Thermo Scientific Chemicals
Description
Silver(I) fluoride is used as a fluorination and desilylation reagent in organic synthesis. In medical filed, it is used as topical caries in aqueous form in dentistry. It is employed in organofluorine chemistry for addition of fluoride across multiple bonds, especially to prepare perfluoroalkylsilver(I) by reacting with alkenes. Further, it is involved in the preparation of tetralkylammonium fluorides by the reaction with tetralkylammonium bromide. In addition to this, it is used as a desulfuration-fluorination reagent on thiourea derived substrates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | ca 1150°C |
Melting Point | 435°C |
Color | White |
Physical Form | Crystalline |
Quantity | 25 g |
Assay Percent Range | 98% |
Solubility Information | Soluble in HF,ammonia,CH3CNSoluble in water,hydrogen fluoride and methanol. |
Formula Weight | 126.87 |
Sensitivity | Hygroscopic; Light and moisture sensitive |
Appearance | Orange-brown crystals |
Show More |
Safety and Handling
Hazard Category | H314-H335 |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | DOT Class: 8, Packaging Group: II |
EINECSNumber | 231-895-8 |
RTECSNumber | VW4250000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.