Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) hydrogenfluoride, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Plastic bottle
12249-52-4
HAgF2
MFCD00042145
silver hydron fluoride,silver salt, h, fluoride,silver 1+ ion hydrogen fluoride,hydrogen . silver 1+ fluoride
12249-52-4
HAgF2
MFCD00042145
silver hydron fluoride,silver salt, h, fluoride,silver 1+ ion hydrogen fluoride,hydrogen . silver 1+ fluoride
Formula Weight 146.87
Physical Form Crystals
Percent Purity Ratio AgF/HF approximately 1:1.06
Packaging Plastic bottle
Color White
Quantity 5g
Chemical Name or Material Silver(I) hydrogenfluoride

GHS H Statement:
Causes severe skin burns and eye damage.
Reacts violently with water.
Contact with acids liberates very toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger