Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) oxide, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
20667-12-3
Ag2O
231.74
MFCD00003404
20667-12-3
Ag2O
231.74
MFCD00003404
Oxidation reagent
Aluminum (Al) 10ppm max.
Assay Percent Range 99+%
Linear Formula Ag2O
Fieser 01,1011; 02,368; 03,252; 04,430; 05,583; 06,515; 07,321; 08,442; 10,350; 11,468
Merck Index 15, 8660
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in dil.hno3 and ammonia,practically insoluble in alcohol
Formula Weight 231.74
Physical Form Powder
Percent Purity 99+%
Elemental Analysis 99.0% min.
Iron (Fe) 30ppm max.
Nitrate 50ppm max.
Packaging Glass bottle
Sulfate 100ppm max.
Color Black
Quantity 250g
Chemical Name or Material Silver(I) oxide
Show More Show Less

GHS H Statement
Causes serious eye damage.
May cause fire or explosion; strong oxidizer.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 243-957-1