Silver(I) oxide, >99% (metals basis) , Alfa Aesar™

Manufacturer: Alfa Aesar 1140736

Quantity 500g

 View more versions of this product

Catalog No. AA1140736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20667-12-3
    Physical Form Powder
    Assay Percent Range >99% (metals basis)
    Molecular Formula Ag2O
    Formula Weight 231.74
    MDL Number MFCD00003404
    Chemical Name or Material Silver(I) oxide

    Specifications

    Formula Weight 231.74
    Quantity 500g
    EINECS Number 243-957-1
    Physical Form Powder
    Density 7.2g/mL
    Assay Percent Range >99% (metals basis)
    Molecular Formula Ag2O
    MDL Number MFCD00003404
    CAS 20667-12-3
    Chemical Name or Material Silver(I) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit