Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver(I) oxide, >99% (metals basis) , Alfa Aesar™

Click to view available options
Quantity:
25g
100g
500g
20667-12-3
Ag2O
MFCD00003404
20667-12-3
Ag2O
MFCD00003404
Assay Percent Range >99% (metals basis)
Solubility Information Soluble in dilute HNO3 and ammonia. Practically insoluble in water. Insoluble in alcohol
Formula Weight 231.74
Physical Form Powder
Density 7.2 g/mL
Sensitivity Light sensitive
Melting Point 230°C (decomposition)
Quantity 25g
Chemical Name or Material Silver(I) oxide

P210-P220-P221-P280-P283-P305+P351+P338-P306+P360-P310-P370+P378q-P371+P380+P375-P501c

H271-H318

DOTInformation : Hazard Class: 5.1; Packaging Group: I

EINECSNumber : 243-957-1

RTECSNumber : VW4900000

TSCA : Yes

Recommended Storage : Ambient temperatures