Alfa Aesar™ Silver(I) oxide, Electrical Grade

For battery plates, pollution control filters

Manufacturer: Alfa Aesar™ 43268A1

Quantity 1kg

 View more versions of this product

Catalog No. AA43268A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20667-12-3
    Molecular Formula Ag2O
    Formula Weight 231.74
    MDL Number MFCD00003404
    UN Number UN1479
    Merck Index 14,8521
    Chemical Name or Material Silver (I) oxide
    Grade Electrical Grade

    Specifications

    Formula Weight 231.74
    Health Hazard 3 GHS P Statement
    P221-P283-P210-P305+P351+P338-P306+P360-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Merck Index 14,8521
    Mesh Size −325 Mesh
    Sensitivity Light Sensitive
    CAS 20667-12-3
    Color Dark brown
    Solubility Information Soluble in dilute HNO3 and ammonia; Practically insoluble in water; Insoluble in alcohol
    Health Hazard 1 Danger
    Density 7.2g/mL
    RTECS Number VW4900000
    Molecular Formula Ag2O
    MDL Number MFCD00003404
    Melting Point 230°C (dec.)
    Health Hazard 2 GHS H Statement
    H271-H318
    May cause fire or explosion
    strong oxidizer.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    TSCA TSCA
    Physical Form Powder
    UN Number UN1479
    Quantity 1kg
    Chemical Name or Material Silver (I) oxide
    Grade Electrical Grade

    GHS H Statement
    H271-H318
    May cause fire or explosion
    strong oxidizer.
    Causes serious eye damage.

    GHS P Statement
    P221-P283-P210-P305+P351+P338-P306+P360-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit