Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver iodate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 4010922

Quantity 100g

 View more versions of this product

Catalog No. AA4010922


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7783-97-3
99%
282.77
Silver iodate
Powder
AgIO3
MFCD00014150

Specifications

7783-97-3
99%
White Powder
MFCD00014150
White
Silver iodate
Powder
AgIO3
282.77
Odorless
100g

Safety and Handling

SDS & Safety Documents

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: II

EINECSNumber : 232-039-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit