Silver iodate, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 4010914

Quantity 25g

 View more versions of this product

Catalog No. AA4010914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-97-3
    Physical Form Powder
    Assay Percent Range 99%
    Molecular Formula AgIO3
    Formula Weight 282.77
    MDL Number MFCD00014150
    Chemical Name or Material Silver iodate

    Specifications

    Physical Form Powder
    Formula Weight 282.77
    Quantity 25g
    Color White
    DOT Information DOT Class: 5.1, Packaging Group: II
    Density 5.53g/cm3 at 20°C
    EINECS Number 232-039-6
    Assay Percent Range 99%
    Molecular Formula AgIO3
    Appearance White Powder
    MDL Number MFCD00014150
    CAS 7783-97-3
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Odor Odorless
    Chemical Name or Material Silver iodate

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit