Silver iodide, 99.9% (metals basis) , Alfa Aesar™

Manufacturer: Alfa Aesar 1141914

Quantity 25g

 View more versions of this product

Catalog No. AA1141914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-96-2
    Physical Form Crystalline
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula AgI
    Formula Weight 234.77
    MDL Number MFCD00003412
    Chemical Name or Material Silver iodide

    Specifications

    Formula Weight 234.77
    Quantity 25g
    EINECS Number 232-038-0
    Physical Form Crystalline
    Density 5.67g/mL
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula AgI
    MDL Number MFCD00003412
    CAS 7783-96-2
    Chemical Name or Material Silver iodide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit