Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver iodide, 99%, pure, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Assay Percent Range | 98.5% min. (ex Ag) |
Linear Formula | AgI |
Merck Index | 15, 8655 |
Solubility Information | Solubility in water: 0.03mg/l |
Formula Weight | 234.77 |
Physical Form | Granular Powder |
Percent Purity | 99% |
Grade | Pure |
Density | 5.6800g/mL |
Packaging | Glass bottle |
Show More |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 232-038-