Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Iodide (Laboratory), Fisher Chemical

Click to view available options
Quantity:
100g
Packaging:
Glass Bottle
7783-96-2
AgI
234.773
MFCD00003412
MSFPLIAKTHOCQP-UHFFFAOYSA-M
silver iodide,neosiluol,neosilvol,silver monoiodide,silver 1+ iodide,colloidal silver iodide,iodanylsilver,silver iodide metals basis,ccris 8490
24563
iodosilver
[Ag]I
7783-96-2
AgI
234.773
MFCD00003412
MSFPLIAKTHOCQP-UHFFFAOYSA-M
silver iodide,neosiluol,neosilvol,silver monoiodide,silver 1+ iodide,colloidal silver iodide,iodanylsilver,silver iodide metals basis,ccris 8490
24563
iodosilver
[Ag]I
Formula Weight 234.77
Physical Form Solid
Grade Laboratory
Identification Pass Test
Packaging Glass Bottle
Color Yellow
Melting Point 552.2°C
Quantity 100g
Chemical Name or Material Silver Iodide