Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver iodide, Premion™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1211122

Quantity 100g

 View more versions of this product

Catalog No. AA1211122


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7783-96-2
MFCD00003412
14,8516
Premion™
234.77
UN3077
Silver iodide

Specifications

7783-96-2
234.77
UN3077
5.67g/mL
Light Sensitive
558°C
Silver iodide
Premion™
Powder
MFCD00003412
14,8516
Odorless
Yellow
100g
99.999%, (metals basis)

Safety and Handling

SDS & Safety Documents

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : VW4450000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit