Alfa Aesar™ Silver metavanadate

Manufacturer: Alfa Aesar™ 4482814

Quantity 25g

 View more versions of this product

Catalog No. AA4482814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13497-94-4
    Molecular Formula AgO3V
    Linear Formula AgVO3
    Formula Weight 206.81
    MDL Number MFCD00041760
    UN Number UN3285
    Synonym Silver vanadium trioxide
    Chemical Name or Material Silver metavanadate

    Specifications

    Formula Weight 206.81
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Linear Formula AgVO3
    Color Red
    Quantity 25g
    Health Hazard 1 Danger
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00041760
    CAS 13497-94-4
    UN Number UN3285
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Sensitivity Light Sensitive
    Molecular Formula AgO3V
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
    Odor Odorless
    Synonym Silver vanadium trioxide
    Chemical Name or Material Silver metavanadate

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit