Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver nanoparticles, 30nm, 0.02mg/mL, supplied in 2mM sodium citrate, 410nm absorption

Click to view available options
Quantity:
25mL
100mL
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.868
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber
23954
CHEBI:9141
silver
[Ag]
Quantity 100mL
Physical Form Liquid
Particle Size 30nm
Sensitivity Light Sensitive
Color Yellow
Chemical Name or Material Silver nanoparticles, 0.02mg/mL, supplied in 2mM sodium citrate, 410nm absorption

storageNote1 : 2° to 8°C