Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

For Chloride Titrations
SDP

Catalog Number 69505
Click to view available options
Quantity:
500 mL
1 L
20 L
Packaging:
Amber Poly Bottle
Cubitainer
7761-88-8, 7732-18-5
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
7761-88-8, 7732-18-5
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O

For Chloride Titrations

Color Colorless
CAS Min % 98.0, 0.47
CAS Max % 100.0, 0.49
Packaging Cubitainer
Solvent or Matrix 1 mL = 1 mg Cl
Quantity 20 L
Physical Form Liquid
Concentration or Composition Notes 0.0282Normal
Chemical Name or Material Silver Nitrate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit