Silver Nitrate, 0.100 Normal (N/10), Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner R70000004X4B

Packaging Black poly
Quantity 16L

 View more versions of this product

Catalog No. 70-001-CS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7761-88-8
    CAS Min % 96.0,1.64
    CAS Max % 100.0,1.7
    Physical Form Liquid
    Chemical Name or Material Silver Nitrate

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Quantity 16L
    Shelf Life 18 Months
    Packaging Black poly
    CAS 7732-18-5,7761-88-8
    Concentration or Composition (by Analyte or Components) 0.100 Normal (N/10)
    CAS Min % 96.0,1.64
    CAS Max % 100.0,1.7
    Chemical Name or Material Silver Nitrate

    18 Months

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit