Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.100 Normal (N/10), Ricca Chemical

Click to view available options
Quantity:
1L
12L
Packaging:
Amber poly
Black poly
7761-88-8, 7732-18-5
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
7761-88-8, 7732-18-5
AgNO3
169.87
MFCD00003414
SQGYOTSLMSWVJD-UHFFFAOYSA-N
silver nitrate, silvernitrate, lunar caustic, silbernitrat, argenti nitras, nitrate d'argent, nitric acid silver i salt, silver mononitrate, silver i nitrate, argerol
24470
CHEBI:32130
silver(1+) nitrate
[Ag+].[O-][N+]([O-])=O
Concentration or Composition (by Analyte or Components) 0.100 Normal (N/10)
CAS Min % 96.0, 1.64
CAS Max % 100.0, 1.7
Physical Form Liquid
Packaging Black poly
Color Colorless
Quantity 12L
Chemical Name or Material Silver Nitrate

ShelfLife : 18 Months