Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.100 Normal USP Testing Method, Reagents

Catalog No. 01336373
Click to view available options
Quantity:
1 L
20 L

Catalog No. 01-336-373

Supplier: Reagents Holdings Llc C730310020F

Only null left
Add to Cart
Add to Cart

Silver Nitrate, 0.100 Normal USP Testing Method, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 20 L
Grade USP
Packaging Cubitainer
Specific Gravity 1.0145
Chemical Name or Material Silver Nitrate, 0.100 Normal USP Testing Method
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.