Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.100N in Denatured Alcohol, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 70281

Packaging Amber Poly
Quantity 4L

 View more versions of this product

Catalog No. 7028-1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

64-17-5,7761-88-8,64-17-5 (Ethyl Alcohol (Ethanol))
100.0,2.12
Silver Nitrate
96.0,2.04
Liquid

Specifications

64-17-5,7761-88-8,64-17-5 (Ethyl Alcohol (Ethanol))
100.0,2.12
Amber Poly
Denatured Alcohol
4L
96.0,2.04
Liquid
0.100Normal
Clear colorless to slight tan
Silver Nitrate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit