Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.282 Normal, Reagents

Catalog No. 01333588
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-333-588 500 mL Glass Bottle
01-333-593 10 L Cubitainer
01-333-592 1 L Glass Bottle
01-333-591 20 L Cubitainer
01-333-590 4 L Glass Bottle
01-333-589 4 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-333-588 Supplier Reagents Holdings Llc Supplier No. CS150800500C
Only null left
Add to Cart
Add to Cart

Silver Nitrate, 0.282 Normal 1 mL = 10 mg Cl, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7761-88-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Nitric Acid silver salt
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
pH 6
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.0401
Chemical Name or Material Silver Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.