Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate, 0.6 N, Unstandardized, Reagents

Catalog No. 01333923
Click to view available options
Quantity:
10 L
15 gal.

Catalog No. 01-333-923

Supplier: Reagents Holdings Llc CS02090015E

Only null left
Add to Cart
Add to Cart
Request a Quote

Silver Nitrate, 0.6 N, Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Silver Nitrate, 0.6 N, Unstandardized
Boiling Point 100°C
CAS 7732-18-5, 7761-88-8
Color Colorless
Physical Form Liquid
Quantity 15 gal.
Molecular Formula AgNO3/H2O
Synonym Nitric Acid silver salt
Grade Reagent
Packaging Poly Drum
Specific Gravity 1.079
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.