Silver nitrate, 99.9995% (metals basis), Alfa Aesar™, Premion™

Manufacturer: Alfa Aesar 1085814

Quantity 25g

 View more versions of this product

Catalog No. AA1085814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7761-88-8
    Physical Form Powder
    Assay Percent Range 99.9995% (metals basis)
    Molecular Formula AgNO3
    Formula Weight 169.87
    MDL Number MFCD00003414
    Chemical Name or Material Silver nitrate

    Specifications

    Formula Weight 169.87
    Quantity 25g
    DOT Information DOT Class: 5.1, Packaging Group: I
    Density 4.352g/cm3 at 20°C
    EINECS Number 231-853-9
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.9995% (metals basis)
    Molecular Formula AgNO3
    Appearance White crystals
    MDL Number MFCD00003414
    CAS 7761-88-8
    Health Hazard 1 H272-H314-H290-H400-H410: May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Silver nitrate

    H272-H314-H290-H400-H410: May intensify fire; oxidizer. Causes severe skin burns and eye damage. May be corrosive to metals. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit