Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver Nitrate, Crystal, USP, 99.8-100.5%, Spectrum™ Chemical

Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Silver Nitrate, Crystal, USP is used as an antibiotic for indolent wounds, corn and callus removal and as a Gonorrheal ophthalmia neonatorum prophylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 1 |
Quantity | 25 g |
Percent Purity | 99.8 to 100.5% |
Grade | USP |
Packaging | Amber Glass Bottle |