Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Nitrate Standard Solution, 1mL = 1mg Chloride, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

7761-88-8
AgNO3
169.872
SQGYOTSLMSWVJD-UHFFFAOYSA-N
24470
CHEBI:32130

High quality chemicals for laboratory and specialized industrial use

CAS 7761-88-8
Molecular Formula AgNO3
InChI Key SQGYOTSLMSWVJD-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[Ag+]
IUPAC Name silver;nitrate
Molecular Weight (g/mol) 169.872
PubChem CID 24470
ChEBI CHEBI:32130
Formula Weight 169.87
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L
Specific Gravity 1L = 1.00kg
Chemical Name or Material Silver Nitrate Standard Solution, 1mL = 1mg Chloride
Grade BAKER ANALYZED™ Reagent
Show More Show Less