Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver perchlorate hydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
14242-05-8
AgClO4·xH2O
207.32
MFCD00149128
IXFNJWPLHOWEPT-UHFFFAOYSA-M
silver perchlorate hydrate,silver perchlorate monohydrate,silver perchlorite,silver perchloric acid,acmc-1brvv,ag.clo4.h2o,silver i perchlorate monohydrate,silver 1+ ion hydrate perchlorate,silver 1+ hydrate perchlorate
15959344
silver;perchlorate;hydrate
O.[O-]Cl(=O)(=O)=O.[Ag+]
14242-05-8
AgClO4·xH2O
207.32
MFCD00149128
IXFNJWPLHOWEPT-UHFFFAOYSA-M
silver perchlorate hydrate,silver perchlorate monohydrate,silver perchlorite,silver perchloric acid,acmc-1brvv,ag.clo4.h2o,silver i perchlorate monohydrate,silver 1+ ion hydrate perchlorate,silver 1+ hydrate perchlorate
15959344
silver;perchlorate;hydrate
O.[O-]Cl(=O)(=O)=O.[Ag+]
Oxidation reagent
Assay Percent Range 98.5% min. (on dry substance) (Argentometry)
Linear Formula AgClO4·xH2O
Merck Index 10, 8358
Solubility Information Solubility in water: 557g/100mL. Other solubilities: soluble in many organic solvents
Formula Weight 207.32
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Chloride 10ppm max
Chlorine (Cl) 10ppm max.
Insoluble in Water 0.005% max.
Insoluble Matter 0.005% max. (in water)
Iron (Fe) 20ppm max.
Packaging Glass bottle
Sulfate 10ppm max.
Water 10% max. (K.F.)
Color White
Quantity 25g
Chemical Name or Material Silver perchlorate hydrate
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger