Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Silver perchlorate monohydrate, 99.9% (metals basis)

Click to view available options
Quantity:
2g
10g
50g
250g
14242-05-8
AgClH2O5
225.329
MFCD00149128
IXFNJWPLHOWEPT-UHFFFAOYSA-M
silver perchlorate hydrate,silver perchlorate monohydrate,silver perchlorite,silver perchloric acid,acmc-1brvv,ag.clo4.h2o,silver i perchlorate monohydrate,silver 1+ ion hydrate perchlorate,silver 1+ hydrate perchlorate
15959344
silver;perchlorate;hydrate
O.[O-]Cl(=O)(=O)=O.[Ag+]
14242-05-8
AgClH2O5
225.329
MFCD00149128
IXFNJWPLHOWEPT-UHFFFAOYSA-M
silver perchlorate hydrate,silver perchlorate monohydrate,silver perchlorite,silver perchloric acid,acmc-1brvv,ag.clo4.h2o,silver i perchlorate monohydrate,silver 1+ ion hydrate perchlorate,silver 1+ hydrate perchlorate
15959344
silver;perchlorate;hydrate
O.[O-]Cl(=O)(=O)=O.[Ag+]
UN Number UN1481
Merck Index 14,8523
Solubility Information Soluble in water and organic solvents.
Formula Weight 225.34 (207.32 Anhydrous)
Physical Form Crystalline
Percent Purity 99.9%
Density 2.800 g/mL
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Color White
Melting Point 485°C (decomposition)
Quantity 250g
Chemical Name or Material Silver perchlorate monohydrate
Assay (metals basis)
Show More Show Less

GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H272-H314

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 232-035-4

TSCA : Yes

Recommended Storage : Ambient temperatures