Alfa Aesar™ Silver perrhenate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4448006

Quantity 5g

 View more versions of this product

Catalog No. AA4448006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20654-56-2
    Molecular Formula AgO4Re
    Linear Formula AgReO4
    Formula Weight 358.1
    MDL Number MFCD00014144
    UN Number UN1479
    Chemical Name or Material Silver perrhenate

    Specifications

    Formula Weight 358.1
    Health Hazard 3 GHS P Statement
    P221-P280g-P273-P305+P351+P338
    Take any precaution to avoid mixing with combustibles.
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    CAS 20654-56-2
    Molecular Formula AgO4Re
    Linear Formula AgReO4
    UN Number UN1479
    Color White
    Sensitivity Light Sensitive
    Quantity 5g
    Density 7.05g/mL
    Physical Form Powder
    MDL Number MFCD00014144
    Melting Point 430°C
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Chemical Name or Material Silver perrhenate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P280g-P273-P305+P351+P338
    Take any precaution to avoid mixing with combustibles.
    Wear protective gloves.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit