Silver plating solution, metal content »28.7g/L, Alfa Aesar™

Manufacturer: Alfa Aesar 44067AK

Quantity 250mL

 View more versions of this product

Catalog No. AA44067AK

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    MDL Number MFCD00003397
    Chemical Name or Material Silver plating solution, Metal content ≈28.7g/L

    Specifications

    Quantity 250mL
    Color Colorless
    DOT Information DOT Class: 6.1, Packaging Group: II
    Physical Form Liquid
    Appearance Colourless liquid
    MDL Number MFCD00003397
    Health Hazard 1 H300-H310-H330-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
    Chemical Name or Material Silver plating solution, Metal content ≈28.7g/L

    H300-H310-H330-H400-H410: Fatal if swallowed. Fatal in contact with skin. Fatal if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit