Learn More
Silver potassium cyanide, ACROS Organics™
Specifications
Merck Index | 15, 7789 |
Formula Weight | 199 |
Physical Form | Crystalline Powder |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Silver (Ag) | 54% min. |
Total Trace Metal Impurities | 0.1% max. |
Color | White |
Quantity | 50g |
Chemical Name or Material | Silver potassium cyanide |
Safety and Handling
GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Contact with acids liberates very toxic gas.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger