Alfa Aesar™ Silver powder, -22 mesh, Premion™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1075606

Quantity 5g

 View more versions of this product

Catalog No. AA1075606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-22-4
    MDL Number MFCD00003397
    UN Number UN3077
    Chemical Name or Material Silver powder
    Grade Premion™

    Specifications

    Mesh Size −22 Mesh
    Odor Characteristic
    Physical Form Powder
    MDL Number MFCD00003397
    UN Number UN3077
    TSCA TSCA
    CAS 7440-22-4
    Quantity 5g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Silver powder
    Assay 99.999%, (metals basis)
    Grade Premion™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit