Alfa Aesar™ Silver powder, spherical, -635 mesh, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4160018

Quantity 50g

 View more versions of this product

Catalog No. AA4160018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-22-4
    MDL Number MFCD00003397
    UN Number UN3077
    Chemical Name or Material Silver powder

    Specifications

    Physical Form Sperical Powder
    Mesh Size −635 Mesh
    MDL Number MFCD00003397
    UN Number UN3077
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    TSCA TSCA
    Odor Characteristic
    CAS 7440-22-4
    Quantity 50g
    Chemical Name or Material Silver powder
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit