Alfa Aesar™ Silver Protein for Histology, Strong

Manufacturer: Alfa Aesar™ 4397014

 View more versions of this product

Catalog No. AA4397014

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Format Powder
    For Use With (Application) Histological stain for microscopic analysis
    CAS 9008-42-8
    Synonym Protargol
    Sensitivity Light Sensitive and Hygroscopic
    Product Type Silver Protein
    Quantity 25g
    Color Pale yellow to brown

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit