Learn More
Alfa Aesar™ Silver Protein for Histology, Strong
Manufacturer:  Alfa Aesar™ 4397006
 View more versions of this product
Catalog No. AA4397006
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5g
Powder
9008-42-8
Light Sensitive and Hygroscopic
Pale yellow to brown
Histological stain for microscopic analysis
Protargol
Silver Protein