Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silver Protein, Mild, 19-23%, Spectrum™

$176.30 - $9760.00

Specifications

Concentration 19 to 23%
CAS 9015-51-4
Color Brown, dark brown or almost black scales or granules
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
18-613-213
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
SI1201KGBL
1 kg
Each for $2,250.00
Add to cart
 
18-613-214
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
SI120100GM
100 g
Each for $564.50
Add to cart
 
18-613-215
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
SI12025GM
25 g
Each for $176.30
Add to cart
 
18-613-216
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
SI1205KGBL
5 kg
Each for $9,760.00
Add to cart
 
18-613-217
View Documents
Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner
SI120500GM
500 g
Each for $1,792.00
Add to cart
 
Description

Description

Specifications

Specifications

19 to 23%
9015-51-4
Brown, dark brown or almost black scales or granules
SDS
Product Certifications

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit