Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silver, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Description
EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.
Specifications
Concentration | 990 to 1010ppm |
Packaging | Glass Bottle |
Quantity | 100 mL |
Grade | Certified |
Vapor Pressure | 14mmHg at 20°C |
Density | 1.0g/cm³ |
Melting Point | 0°C |
Boiling Point | 100°C |
Physical Form | Liquid |
Chemical Name or Material | Silver, Reference Standard Solution, 1000ppm ±1% |
Safety and Handling
DOTInformation | DOT Class 8, : Corrosive |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction